Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039259.1 | NP_001034348.1 | 8 | 1308 | ||
XM_006248447.2 | XP_006248509.1 | 8 | 1310 | |||
XM_008768710.1 | XP_008766932.1 | 8 | 1316 | |||
GenBank mRNA | AY886535.1 | - | 8 | 1308 | ||
BC127441.1 | - | 8 | 1310 | 7111596 |