Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100501.1 | NP_001093971.1 | 16 | 2306 | ||
XM_006248015.2 | XP_006248077.1 | 16 | 2322 | |||
GenBank mRNA | BC087705.1 | - | 15 | 2170 | ||
BI282215.1 | - | 3 | 594 | |||
FQ221582.1 | - | 15 | 3177 | |||
FQ226796.1 | - | 3 | 772 |