Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105916.1 | NP_001099386.1 | 15 | 1755 | ||
XM_006249116.2 | XP_006249178.1 | 14 | 1707 | |||
XM_006249117.1 | XP_006249179.1 | 15 | 1723 | |||
XM_006249118.1 | XP_006249180.1 | 15 | 1726 | |||
XM_008769125.1 | XP_008767347.1 | 14 | 1836 | |||
XM_008769126.1 | XP_008767348.1 | 14 | 1666 | |||
XM_008769127.1 | XP_008767349.1 | 14 | 1707 | |||
XM_008769128.1 | XP_008767350.1 | 14 | 1707 | |||
GenBank mRNA | BC166451.1 | - | 14 | 1820 | 8371649 |