Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563293 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769809.1 | XP_008768031.1 | 1 | 1234 | ||
XM_222933.3 | XP_222933.2 | 1 | 1234 |