Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277600.1 | NP_001264529.1 | 3 | 378 | ||
NM_001277601.1 | NP_001264530.1 | 3 | 378 | |||
NM_001277602.1 | NP_001264531.1 | 2 | 352 | |||
XM_006251858.2 | XP_006251920.1 | 2 | 846 | |||
XM_006251859.2 | XP_006251921.1 | 2 | 339 | |||
XM_006251861.2 | XP_006251923.1 | 2 | 845 | |||
XM_006251862.2 | XP_006251924.1 | 4 | 529 | |||
XM_006251863.2 | XP_006251925.1 | 2 | 339 | |||
XM_006251864.2 | XP_006251926.1 | 4 | 434 | |||
XM_006251865.2 | XP_006251927.1 | 2 | 845 | |||
XM_006251866.1 | XP_006251928.1 | 3 | 365 | |||
XM_008770508.1 | XP_008768730.1 | 3 | 873 | |||
XM_008770509.1 | XP_008768731.1 | 3 | 1941 | |||
XM_008770510.1 | XP_008768732.1 | 3 | 373 | |||
XM_008770511.1 | XP_008768733.1 | 3 | 373 | |||
XM_008770512.1 | XP_008768734.1 | 3 | 365 | |||
GenBank mRNA | BC097346.1 | - | 2 | 302 | 7442283 | |
CB582273.1 | - | 3 | 328 | |||
FQ222540.1 | - | 1 | 28 | |||
FQ227266.1 | - | 1 | 28 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271241.1 | NP_001258170.1 | 3 | 343 | ||
XM_003751394.2 | XP_003751442.1 | 3 | 329 | |||
XM_003751395.2 | XP_003751443.1 | 3 | 329 | |||
XM_006251449.2 | XP_006251511.1 | 3 | 1940 | |||
XM_006251450.2 | XP_006251512.1 | 2 | 844 | |||
XM_006251451.1 | XP_006251513.1 | 2 | 303 | |||
XM_006251452.2 | XP_006251514.1 | 3 | 329 | |||
XM_006251453.1 | XP_006251515.1 | 4 | 493 | |||
XM_006251454.1 | XP_006251516.1 | 4 | 398 | |||
XM_006251455.2 | XP_006251517.1 | 2 | 843 | |||
XM_006251456.1 | XP_006251518.1 | 2 | 303 | |||
XM_006251457.1 | XP_006251519.1 | 2 | 303 | |||
XM_006251458.2 | XP_006251520.1 | 3 | 329 | |||
XM_008770242.1 | XP_008768464.1 | 3 | 837 | |||
XM_008770243.1 | XP_008768465.1 | 3 | 329 | |||
GenBank mRNA | AY724499.1 | - | 2 | 771 | ||
FQ214198.1 | - | 1 | 28 | |||
FQ215781.1 | - | 1 | 28 | |||
FQ227893.1 | - | 1 | 28 | |||
FQ228082.1 | - | 1 | 28 |