Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277600.1 | NP_001264529.1 | 5, 6 | 614 | ||
NM_001277601.1 | NP_001264530.1 | 5, 6 | 575 | |||
NM_001277602.1 | NP_001264531.1 | 4, 5 | 549 | |||
XM_006251858.2 | XP_006251920.1 | 4, 5 | 1082 | |||
XM_006251859.2 | XP_006251921.1 | 4, 5 | 575 | |||
XM_006251861.2 | XP_006251923.1 | 4, 5 | 1042 | |||
XM_006251862.2 | XP_006251924.1 | 6, 7 | 726 | |||
XM_006251863.2 | XP_006251925.1 | 4, 5 | 536 | |||
XM_006251864.2 | XP_006251926.1 | 6, 7 | 631 | |||
XM_006251865.2 | XP_006251927.1 | 4, 5 | 1042 | |||
XM_006251866.1 | XP_006251928.1 | 5, 6 | 562 | |||
XM_008770508.1 | XP_008768730.1 | 5, 6 | 1109 | |||
XM_008770509.1 | XP_008768731.1 | 5, 6 | 2177 | |||
XM_008770510.1 | XP_008768732.1 | 5, 6 | 609 | |||
XM_008770511.1 | XP_008768733.1 | 5, 6 | 609 | |||
XM_008770512.1 | XP_008768734.1 | 5, 6 | 562 | |||
GenBank mRNA | BC097346.1 | - | 4, 5 | 499 | 7442283 | |
CB582273.1 | - | 5, 6 | 564 | |||
FQ214011.1 | - | 3, 4 | 186 | |||
FQ227266.1 | - | 3, 4 | 225 |
Gene Symbol: LOC100911576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271241.1 | NP_001258170.1 | 5, 6 | 540 | ||
XM_003751394.2 | XP_003751442.1 | 5, 6 | 526 | |||
XM_003751395.2 | XP_003751443.1 | 5, 6 | 565 | |||
XM_006251449.2 | XP_006251511.1 | 5, 6 | 2176 | |||
XM_006251450.2 | XP_006251512.1 | 4, 5 | 1080 | |||
XM_006251451.1 | XP_006251513.1 | 4, 5 | 539 | |||
XM_006251452.2 | XP_006251514.1 | 5, 6 | 565 | |||
XM_006251453.1 | XP_006251515.1 | 6, 7 | 690 | |||
XM_006251454.1 | XP_006251516.1 | 6, 7 | 595 | |||
XM_006251455.2 | XP_006251517.1 | 4, 5 | 1040 | |||
XM_006251456.1 | XP_006251518.1 | 4, 5 | 500 | |||
XM_006251457.1 | XP_006251519.1 | 4, 5 | 500 | |||
XM_006251458.2 | XP_006251520.1 | 5, 6 | 526 | |||
XM_008770242.1 | XP_008768464.1 | 5, 6 | 1073 | |||
XM_008770243.1 | XP_008768465.1 | 5, 6 | 565 | |||
GenBank mRNA | AY724499.1 | - | 4, 5 | 968 | ||
FQ214198.1 | - | 3, 4 | 225 | |||
FQ215781.1 | - | 3, 4 | 264 | |||
FQ227893.1 | - | 3, 4 | 225 | |||
FQ228082.1 | - | 3, 4 | 264 |