Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tssk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271325.1 | NP_001258254.1 | 1 | 294 | ||
XM_006251938.2 | XP_006252000.1 | 1 | 6748 | |||
XM_006251939.2 | XP_006252001.1 | 1 | 6748 | |||
XM_008770674.1 | XP_008768896.1 | 2 | 6682 | |||
GenBank mRNA | BC079202.1 | - | 1 | 319 | 7131733 |