Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100944.1 | NP_001094414.1 | 2 | 553 | ||
XM_008771316.1 | XP_008769538.1 | 6 | 1359 | |||
XM_008771317.1 | XP_008769539.1 | 5 | 1307 | |||
XM_008771318.1 | XP_008769540.1 | 7 | 1413 | |||
XM_008771319.1 | XP_008769541.1 | 6 | 1359 | |||
XM_008771320.1 | XP_008769542.1 | 4 | 1573 | |||
XM_008771321.1 | XP_008769543.1 | 3 | 986 | |||
XM_008771322.1 | XP_008769544.1 | 6 | 1359 | |||
GenBank mRNA | BC128761.1 | - | 2 | 553 | 7113007 |
Gene Symbol: LOC100911229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771345.1 | XP_008769567.1 | 1679 |