Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077640.1 | NP_001071108.1 | 2, 3 | 238 | ||
GenBank mRNA | AB020978.1 | - | 2, 3 | 238 | ||
BC168643.1 | - | 2, 3 | 238 | 9092341 | ||
FQ218856.1 | - | 2, 3 | 256 |