Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169578.1 | NP_001163049.1 | 1 | 128 | ||
XM_006253671.2 | XP_006253733.1 | 2 | 193 | |||
XM_008771489.1 | XP_008769711.1 | 2 | 212 |