Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106135.2 | NP_001099605.2 | 12 | 1289 | ||
XM_006254716.2 | XP_006254778.1 | 12 | 1315 | |||
GenBank mRNA | BC166853.1 | - | 12 | 1264 | 9092757 | |
CO567837.1 | - | 6 | 555 | |||
DV723418.1 | - | 2 | 105 |