Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077667.1 | NP_001071135.1 | 5 | 511 | ||
XM_008772113.1 | XP_008770335.1 | 6 | 805 | |||
XM_008772114.1 | XP_008770336.1 | 5 | 961 | |||
XM_008772115.1 | XP_008770337.1 | 5 | 584 | |||
XM_008772116.1 | XP_008770338.1 | 5 | 590 | |||
GenBank mRNA | BC127517.1 | - | 5 | 511 | 7104038 |