Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106181.1 | NP_001099651.1 | 16 | 1573 | ||
XM_006255479.1 | XP_006255541.1 | 12 | 1731 | |||
GenBank mRNA | BC158728.1 | - | 16 | 1475 | 9091646 |