Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
siRNA ID
RSS354183
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
2
82
XM_008766672.1
XP_008764894.1
2
155
GenBank mRNA
BC060578.1
-
2
100
6919235
D25224.1
-
2
83
FJ386454.1
-
1
63
FQ209463.1
-
2
96
FQ209469.1
-
2
96
FQ209490.1
-
2
97
FQ209667.1
-
2
96
FQ209729.1
-
2
96
FQ211189.1
-
2
96
FQ212660.1
-
2
96
FQ212893.1
-
2
96
FQ213275.1
-
2
96
FQ213317.1
-
2
96
FQ213334.1
-
2
96
FQ213623.1
-
2
99
FQ213835.1
-
2
97
FQ213894.1
-
2
96
FQ215242.1
-
2
96
FQ215292.1
-
2
97
FQ216063.1
-
2
96
FQ216085.1
-
2
96
FQ216130.1
-
2
96
FQ216245.1
-
2
96
FQ216997.1
-
2
96
FQ217664.1
-
2
96
FQ219586.1
-
2
96
FQ219785.1
-
2
99
FQ220012.1
-
2
96
FQ220017.1
-
2
94
FQ220075.1
-
2
97
FQ220084.1
-
2
99
FQ220126.1
-
2
96
FQ220134.1
-
2
96
FQ220171.1
-
2
95
FQ220184.1
-
2
96
FQ220347.1
-
2
98
FQ220461.1
-
2
77
FQ220468.1
-
2
96
FQ220661.1
-
2
99
FQ220904.1
-
2
97
FQ220964.1
-
2
96
FQ221103.1
-
2
96
FQ221334.1
-
2
116
FQ221443.1
-
2
97
FQ221672.1
-
2
96
FQ222093.1
-
2
98
FQ222280.1
-
2
96
FQ222542.1
-
2
98
FQ222757.1
-
2
94
FQ223223.1
-
2
96
FQ224747.1
-
2
96
FQ225510.1
-
2
97
FQ226408.1
-
2
96
FQ227056.1
-
2
96
FQ227600.1
-
2
97
FQ228002.1
-
2
96
FQ228212.1
-
2
97
FQ229290.1
-
2
98
FQ229305.1
-
2
98
FQ229367.1
-
2
97
FQ229516.1
-
2
96
FQ229579.1
-
2
96
FQ229598.1
-
2
97
FQ229712.1
-
2
96
FQ229787.1
-
2
96
FQ229850.1
-
2
98
FQ229859.1
-
2
94
FQ229890.1
-
2
96
FQ229918.1
-
2
116
FQ229945.1
-
2
96
FQ229970.1
-
2
96
FQ230088.1
-
2
96
FQ230106.1
-
2
96
FQ230180.1
-
2
97
FQ230258.1
-
2
96
FQ231622.1
-
2
94
FQ232305.1
-
2
98
FQ232568.1
-
2
97
FQ233068.1
-
2
96
FQ233112.1
-
2
96
FQ233340.1
-
2
98
FQ233547.1
-
2
97
FQ233919.1
-
2
96
FQ234511.1
-
2
96
FQ234742.1
-
2
96
FQ234765.1
-
2
96
FQ234772.1
-
2
96
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-nlqsn:80/100.66.77.47:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0