Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019183.1 | NP_062056.1 | 7 | 1248 | ||
GenBank mRNA | AI603593.1 | - | 1 | 256 | ||
BC127451.1 | - | 7 | 1123 | 7379877 | ||
X80130.1 | - | 7 | 1134 |
Gene Symbol: Acta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X00306.1 | - | 206 |