Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013066.1 | NP_001013084.1 | 4, 5 | 999 | ||
XM_008759130.1 | XP_008757352.1 | 14, 15 | 4423 | |||
XM_008759131.1 | XP_008757353.1 | 14, 15 | 4423 | |||
GenBank mRNA | BC091303.1 | - | 4, 5 | 999 | 7309905 |