Gene Symbol: Ndufc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009290.1 | NP_001009290.1 | 1, 2 | 249 | ||
GenBank mRNA | BC086952.1 | - | 1, 2 | 249 | 7325437 | |
FQ220552.1 | - | 1, 2 | 298 | |||
FQ224080.1 | - | 1, 2 | 294 |