Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134428.1 | NP_001127900.1 | 2 | 113 | ||
NM_001134429.1 | NP_001127901.1 | 2 | 149 | |||
XM_006230756.1 | XP_006230818.1 | 2 | 201 | |||
XM_006230757.1 | XP_006230819.1 | 2 | 165 | |||
XM_006230758.2 | XP_006230820.1 | 2 | 603 | |||
GenBank mRNA | BG669308.1 | - | 2 | 113 | ||
BI285619.1 | - | 5 | 499 | |||
H35224.1 | - | 2 | 103 |