Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134428.1 | NP_001127900.1 | 2, 3 | 261 | ||
NM_001134429.1 | NP_001127901.1 | 2, 3 | 297 | |||
XM_006230756.1 | XP_006230818.1 | 2, 3 | 349 | |||
XM_006230757.1 | XP_006230819.1 | 2, 3 | 313 | |||
XM_006230758.2 | XP_006230820.1 | 2, 3 | 751 | |||
GenBank mRNA | BG669308.1 | - | 2, 3 | 261 | ||
BI285619.1 | - | 4, 5 | 351 | |||
H35224.1 | - | 2, 3 | 250 |