Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106366.1 | NP_001099836.1 | 10, 11 | 1070 | ||
XM_006231476.2 | XP_006231538.1 | 10, 11 | 1816 |