Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps4x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007600.1 | NP_001007601.1 | 1 | 244 | ||
GenBank mRNA | BC086560.1 | - | 1 | 244 | 7303578 | |
FQ220060.1 | - | 1 | 267 | |||
FQ226324.1 | - | 1 | 267 | |||
FQ230280.1 | - | 1 | 267 | |||
X14210.1 | - | 1 | 265 |
Gene Symbol: LOC100362640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730258.3 | XP_002730304.1 | 3 | 332 |