Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024133.2 | NP_077047.1 | 1 | 257 | ||
NM_177982.1 | NP_817091.1 | 1 | 257 | |||
GenBank mRNA | U38370.1 | - | 1 | 272 | ||
U38373.1 | - | 1 | 272 |