Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017212.2 | NP_058908.2 | 7 | 815 | ||
XM_008768268.1 | XP_008766490.1 | 11 | 1988 | |||
XM_008768269.1 | XP_008766491.1 | 10 | 1901 | |||
XM_008768271.1 | XP_008766493.1 | 9 | 1814 | |||
XM_008768272.1 | XP_008766494.1 | 10 | 1789 | |||
XM_008768274.1 | XP_008766496.1 | 10 | 1222 | |||
XM_008768275.1 | XP_008766497.1 | 9 | 1135 | |||
XM_008768277.1 | XP_008766499.1 | 9 | 1021 | |||
XM_008768278.1 | XP_008766500.1 | 8 | 934 | |||
XM_008768282.1 | XP_008766504.1 | 12 | 2043 | |||
GenBank mRNA | CB615367.1 | - | 4 | 478 | ||
M84156.1 | - | 10 | 1646 | |||
X79321.1 | - | 7 | 902 |