Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761059.1 | XP_008759281.1 | 2, 3 | 2744 | ||
XM_215580.6 | XP_215580.2 | 2, 3 | 2726 |
Set Membership: |
![]() |