Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rhoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106461.1 | NP_001099931.1 | 5 | 707 | ||
XM_006233074.1 | XP_006233136.1 | 5 | 681 | |||
GenBank mRNA | BC166849.1 | - | 5 | 723 | 9088252 |