Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021989.2 | NP_068824.1 | 5 | 613 | ||
GenBank mRNA | AJ409332.1 | - | 5 | 613 | ||
BC084714.1 | - | 5 | 705 | 7303706 | ||
L31884.1 | - | 5 | 696 | |||
S72594.1 | - | 5 | 717 | |||
S82718.1 | - | 5 | 706 | |||
U14526.1 | - | 5 | 696 |