Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
siRNA ID
RSS355622
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
3
290
GenBank mRNA
DQ273679.1
-
3
290
FQ215069.1
-
354
FQ216531.1
-
354
FQ216869.1
-
354
FQ216875.1
-
384
FQ216940.1
-
346
FQ216956.1
-
367
FQ217047.1
-
354
FQ217054.1
-
354
FQ217106.1
-
383
FQ217137.1
-
354
FQ217169.1
-
354
FQ217172.1
-
354
FQ217195.1
-
354
FQ217236.1
-
365
FQ217299.1
-
354
FQ217378.1
-
354
FQ217706.1
-
354
FQ217788.1
-
354
FQ217840.1
-
354
FQ217842.1
-
354
FQ217896.1
-
353
FQ217902.1
-
354
FQ217974.1
-
383
FQ217984.1
-
354
FQ218010.1
-
354
FQ218048.1
-
384
FQ218061.1
-
354
FQ223543.1
-
354
FQ223572.1
-
354
FQ223583.1
-
354
FQ223607.1
-
354
FQ223623.1
-
354
FQ223636.1
-
354
FQ223643.1
-
354
FQ223690.1
-
354
FQ223692.1
-
354
FQ223711.1
-
354
FQ223713.1
-
354
FQ223719.1
-
354
FQ223773.1
-
366
FQ223816.1
-
375
FQ223825.1
-
354
FQ223832.1
-
354
FQ223835.1
-
384
FQ223976.1
-
354
FQ223986.1
-
354
FQ224079.1
-
354
FQ224114.1
-
383
FQ224128.1
-
354
FQ224223.1
-
354
FQ224258.1
-
354
FQ224340.1
-
354
FQ224378.1
-
354
FQ224387.1
-
354
FQ224450.1
-
354
FQ224462.1
-
354
FQ224599.1
-
354
FQ224648.1
-
354
FQ224735.1
-
355
FQ224749.1
-
354
FQ224755.1
-
383
FQ224795.1
-
354
FQ224811.1
-
354
FQ224822.1
-
354
FQ224827.1
-
351
J00793.1
-
1
74
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-zl6vn:80/100.66.77.21:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0