Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNC2 SIRNA Assays ›
Gene Symbol:
Tnnc2
Gene Name:
troponin C type 2 (fast)
Gene Aliases:
-
Chromosome Location:
Chr.3: 161243747 - 161244831 on Build Rnor_6.0
UniGene ID:
Rn.43529
Species:
Rat
siRNA ID
RSS355624
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037351.1
NP_001032428.1
2
76
GenBank mRNA
DQ273679.1
-
2
76
FQ215069.1
-
140
FQ216531.1
-
140
FQ216869.1
-
140
FQ216875.1
-
170
FQ216940.1
-
132
FQ216956.1
-
153
FQ217047.1
-
140
FQ217054.1
-
140
FQ217106.1
-
169
FQ217137.1
-
140
FQ217169.1
-
140
FQ217172.1
-
140
FQ217195.1
-
140
FQ217236.1
-
151
FQ217299.1
-
140
FQ217371.1
-
140
FQ217378.1
-
140
FQ217706.1
-
140
FQ217788.1
-
140
FQ217840.1
-
140
FQ217842.1
-
140
FQ217896.1
-
139
FQ217902.1
-
140
FQ217974.1
-
169
FQ217984.1
-
140
FQ218010.1
-
140
FQ218048.1
-
170
FQ218061.1
-
140
FQ223543.1
-
140
FQ223572.1
-
140
FQ223583.1
-
140
FQ223607.1
-
140
FQ223623.1
-
140
FQ223636.1
-
140
FQ223643.1
-
140
FQ223690.1
-
140
FQ223692.1
-
140
FQ223711.1
-
140
FQ223713.1
-
140
FQ223719.1
-
140
FQ223773.1
-
152
FQ223816.1
-
161
FQ223825.1
-
140
FQ223832.1
-
140
FQ223835.1
-
170
FQ223976.1
-
140
FQ224079.1
-
140
FQ224114.1
-
169
FQ224128.1
-
140
FQ224223.1
-
140
FQ224258.1
-
140
FQ224340.1
-
140
FQ224376.1
-
155
FQ224378.1
-
140
FQ224387.1
-
140
FQ224450.1
-
140
FQ224462.1
-
140
FQ224599.1
-
140
FQ224648.1
-
140
FQ224672.1
-
140
FQ224735.1
-
141
FQ224749.1
-
140
FQ224755.1
-
169
FQ224795.1
-
140
FQ224811.1
-
140
FQ224822.1
-
140
FQ224827.1
-
139
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
actin binding
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline