Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017349.2 | NP_059045.2 | 1, 2 | 48 | ||
NM_145717.1 | NP_663769.1 | 2, 3 | 92 | |||
XM_006242911.2 | XP_006242973.1 | 2, 3 | 249 | |||
XM_006242912.2 | XP_006242974.1 | 2, 3 | 197 | |||
GenBank mRNA | AF129400.1 | - | 2, 3 | 92 | ||
AF233060.1 | - | 1, 2 | 48 | |||
X70062.1 | - | 2, 3 | 56 |