Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015003.2 | NP_001015003.1 | 8 | 880 | ||
GenBank mRNA | AY973953.1 | - | 8 | 867 | ||
BC099779.1 | - | 8 | 865 | |||
BC127455.1 | - | 8 | 872 | 7112865 |