Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cdca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007648.1 | NP_001007649.1 | 2 | 627 | ||
XM_006237341.2 | XP_006237403.1 | 2 | 211 | |||
GenBank mRNA | BC079204.1 | - | 2 | 627 | 7131776 | |
FQ235136.1 | - | 2 | 165 |