Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Churc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106741.2 | NP_001100211.1 | 4 | 336 | ||
GenBank mRNA | BC168770.1 | - | 4 | 336 | 6918115 |