Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008516.2 | NP_001008516.2 | 1 | 232 | ||
XM_006240374.2 | XP_006240436.1 | 2 | 694 | |||
XM_006240375.2 | XP_006240437.1 | 2 | 695 | |||
XM_008764759.1 | XP_008762981.1 | 2 | 1930 | |||
GenBank mRNA | AY383731.1 | - | 1 | 232 |