Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013955.1 | NP_001013977.1 | 1 | 296 | ||
XM_008766513.1 | XP_008764735.1 | 1 | 335 | |||
GenBank mRNA | BC087151.1 | - | 1 | 296 | 7131982 |