Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134737.1 | NP_001128209.1 | 5, 6 | 1984 | ||
GenBank mRNA | BC168206.1 | - | 5, 6 | 1029 | 7130974 |