Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037189.1 | NP_001032266.1 | 7 | 1039 | ||
XM_008768018.1 | XP_008766240.1 | 6 | 2187 | |||
GenBank mRNA | BC099120.1 | - | 7 | 1039 | 7462721 |