Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107192.1 | NP_001100662.1 | 3 | 602 | ||
XM_006250112.2 | XP_006250174.1 | 3 | 612 | |||
XM_008769649.1 | XP_008767871.1 | 3 | 705 | |||
GenBank mRNA | AY724478.1 | - | 3 | 573 | ||
FQ231128.1 | - | 3 | 579 |