Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107208.2 | NP_001100678.1 | 329 | |||
GenBank mRNA | BC167769.1 | - | 460 | 7456582 | ||
EU711414.1 | - | 329 | ||||
EU711415.1 | - | 329 | ||||
EU711416.1 | - | 329 | ||||
EU711417.1 | - | 329 |