Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006995.1 | NP_001006996.1 | 5 | 637 | ||
GenBank mRNA | AY325187.1 | - | 611 | |||
BC083872.1 | - | 5 | 637 | 7128564 |
Gene Symbol: LOC102554294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227914.2 | XP_006227976.1 | 2 | 839 |
Gene Symbol: RGD1562948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748697.3 | XP_003748745.2 | 5 | 731 |