Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231909.2 | XP_006231971.1 | 6 | 1071 | ||
XM_008760730.1 | XP_008758952.1 | 3 | 3531 | |||
GenBank mRNA | AY325185.1 | - | 4 | 487 |