Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Accs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267534.1 | NP_001254463.1 | 5 | 526 | ||
XM_006234577.2 | XP_006234639.1 | 5 | 531 | |||
XM_006234578.2 | XP_006234640.1 | 5 | 660 | |||
XM_006234579.2 | XP_006234641.1 | 6 | 603 | |||
XM_006234580.2 | XP_006234642.1 | 5 | 620 | |||
XM_008762028.1 | XP_008760250.1 | 5 | 659 | |||
GenBank mRNA | BC083866.1 | - | 5 | 526 |