Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191752.1 | NP_001178681.1 | 8 | 864 | ||
XM_006237236.2 | XP_006237298.1 | 9 | 1110 | |||
XM_006237237.2 | XP_006237299.1 | 9 | 1098 | |||
XM_006237238.1 | XP_006237300.1 | 10 | 1153 | |||
XM_006237239.1 | XP_006237301.1 | 10 | 1135 | |||
XM_006237240.2 | XP_006237302.1 | 9 | 2021 | |||
XM_006237241.1 | XP_006237303.1 | 9 | 956 | |||
XM_006237242.2 | XP_006237304.1 | 8 | 2041 | |||
XM_006237243.2 | XP_006237305.1 | 8 | 1967 | |||
XM_006237244.2 | XP_006237306.1 | 8 | 864 | |||
XM_008763244.1 | XP_008761466.1 | 9 | 1110 | |||
XM_008763245.1 | XP_008761467.1 | 9 | 2095 | |||
XM_008763246.1 | XP_008761468.1 | 9 | 951 |