Gene Symbol: Nipsnap3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009422.1 | NP_001009422.1 | 2 | 244 | ||
XM_006238164.2 | XP_006238226.1 | 2 | 299 | |||
GenBank mRNA | BC087643.1 | - | 2 | 244 | 7307943 | |
FQ219476.1 | - | 2 | 296 | |||
FQ221422.1 | - | 2 | 213 |