Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108036.2 | NP_001101506.2 | 8 | 1563 | ||
XM_006240262.2 | XP_006240324.1 | 8 | 1444 | |||
XM_006240263.2 | XP_006240325.1 | 8 | 1547 | |||
XM_008764775.1 | XP_008762997.1 | 8 | 1344 | |||
XM_008764776.1 | XP_008762998.1 | 8 | 1444 | |||
XM_008764777.1 | XP_008762999.1 | 8 | 1444 |