Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012118.1 | NP_001012118.1 | 4 | 1252 | ||
XM_006241542.2 | XP_006241604.1 | 4 | 1375 | |||
GenBank mRNA | BC079211.1 | - | 4 | 1252 | 7131819 |