Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127546.1 | NP_001121018.1 | 4 | 586 | ||
XM_008765714.1 | XP_008763936.1 | 4 | 1304 | |||
GenBank mRNA | BC162033.1 | - | 4 | 586 | 9103157 |