Search Thermo Fisher Scientific
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181637.1 | NP_853668.1 | 9 | 915 | ||
XM_006235325.2 | XP_006235387.1 | 10 | 1166 | |||
XM_008762347.1 | XP_008760569.1 | 7 | 1263 | |||
GenBank mRNA | AB086841.1 | - | 9 | 950 | ||
BC097269.1 | - | 10 | 1041 |