Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Asb15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236158.2 | XP_006236220.1 | 8 | 1325 | ||
XM_008762831.1 | XP_008761053.1 | 8 | 1481 | |||
XM_008762832.1 | XP_008761054.1 | 8 | 1282 | |||
XM_008762833.1 | XP_008761055.1 | 8 | 1503 | |||
GenBank mRNA | AY339371.1 | - | 1 | 42 | ||
BC097405.1 | - | 8 | 1037 |