Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012169.1 | NP_001012169.1 | 12, 13 | 1435 | ||
XM_006229994.1 | XP_006230056.1 | 12, 13 | 1495 | |||
GenBank mRNA | BC083578.1 | - | 12, 13 | 1435 | 7127506 |