Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024274.1 | NP_001019445.1 | 283 | |||
GenBank mRNA | AY281367.1 | - | 283 |